Merck
  • Efficient cleavage of DS DNA by bleomycin conjugated via hexaethylene glycol linker to triplex-forming oligonucleotides.
Efficient cleavage of DS DNA by bleomycin conjugated via hexaethylene glycol linker to triplex-forming oligonucleotides.
Nucleosides, nucleotides & nucleic acids (2004-11-25)
Pavel Vorobjev, Olessia Tchaika, Valentina Zarytova
ABSTRACT
New conjugates of bleomycin A5 with oligonucleotides are synthesized. The bleomycin residue was attached to the 3'- or 5'- terminus of hexadecathymidilate via a hexaethylene glycol phosphate linker. Newly designed conjugates were shown to cleave site-specifically both strands of a dsDNA fragment within a triplex. The maximum extent of cleavage for individual strand amounts to 61%.
Sigma-Aldrich
Hexaethylene glycol, 97%