Merck
  • Depudecin: a novel compound inducing the flat phenotype of NIH3T3 cells doubly transformed by ras- and src-oncogene, produced by Alternaria brassicicola.
Depudecin: a novel compound inducing the flat phenotype of NIH3T3 cells doubly transformed by ras- and src-oncogene, produced by Alternaria brassicicola.
The Journal of antibiotics (1992-06-01)
M Matsumoto, S Matsutani, K Sugita, H Yoshida, F Hayashi, Y Terui, H Nakai, N Uotani, Y Kawamura, K Matsumoto
ABSTRACT
A novel compound depudecin inducing the flat phenotype of ras- and src- transformed NIH3T3 cells at a concentration of 1 microgram/ml was isolated from the culture broth of Alternaria brassicicola. Based on its spectroscopic characteristics and X-ray crystallographic analysis of its bis-(1S)-(-)-camphanate, the structure of depudecin was determined to be (2R,3S,4S,5E,7S,8S,9R)-2,9- dihydroxy-3,4;7,8-diepoxy-undeca-5,10-diene.