Merck
  • Lipase-catalyzed kinetic resolution on solid-phase via a "capture and release" strategy.
Lipase-catalyzed kinetic resolution on solid-phase via a "capture and release" strategy.
Journal of the American Chemical Society (2003-11-13)
Cara E Humphrey, Nicholas J Turner, Morag A M Easson, Sabine L Flitsch, Rein V Ulijn
ABSTRACT
The lipase-catalyzed kinetic resolution of (R/S)-3-phenylbutyric acid 2 using solid-supported cyclohexane-1,3-dione (CHD) 6 is described. In each case the predominant enantiomer observed, after cleavage from the resin, was (R)-(-)-3-phenylbutyric acid (R)-2 (ee > 99%) rather than the expected (S)-enantiomer of 2. This observation is in contrast to the fact that Chromobacterium viscosum lipase shows high enantiospecificity for the (S)-enantiomer in the corresponding solution-phase hydrolysis reactions. The (R)-acyl group was subsequently released from the resin by NaOH hydrolysis, and the yield of the reaction could be improved by triple acylation of the resin.
Sigma-Aldrich
Poly(ethylene glycol) bis(amine), Mw 20,000
Sigma-Aldrich
Poly(ethylene glycol) bis(amine), Mw 3,000
Sigma-Aldrich
Poly(ethylene glycol) bis(amine), Mw 2,000
Sigma-Aldrich
Poly(ethylene glycol) bis(amine), average Mn 3400
Sigma-Aldrich
Poly(ethylene glycol) bis(amine), Mw 10,000
Sigma-Aldrich
Poly(ethylene glycol) diamine, average Mn 10,000
Sigma-Aldrich
Poly(ethylene glycol) diamine, average Mn 2,000
Sigma-Aldrich
Poly(ethylene glycol) bis(amine), Mw 6,000
Sigma-Aldrich
Poly(ethylene glycol) diamine, average Mn 6,000
Sigma-Aldrich
Poly(ethylene glycol) diamine, average Mn 3,000