Merck
  • [Plasmids for biodegradation of 2,6-dimethylpyridine, 2,4-dimethylpyridine, and pyridine in strains of Arthrobacter].
[Plasmids for biodegradation of 2,6-dimethylpyridine, 2,4-dimethylpyridine, and pyridine in strains of Arthrobacter].
Molekuliarnaia genetika, mikrobiologiia i virusologiia (1992-05-01)
S R Agapova, A L Andreeva, I I Starovoĭtov, L I Vorob'eva, P B Terent'ev
ABSTRACT
Arthrobacter crysallopoietes strain KM-4 degrading 2,6-dimethylpyridine and strain KM-4a degrading both 2,6-dimethylpyridine and pyridine, Arthrobacter sp. KM-4b degrading 2,4-dimethylpyridine were isolated from soil. Arthrobacter crystallopoietes KM-4 and Arthrobacter sp. KM-4b contain 100 Md plasmids pBS320 and pBS323. Arthrobacter crystallopietes KM-4a harbours a 100 Md and 80 Md plasmids. Plasmid curing and conjugation transfer results confirm that these plasmids are involved in degradation of 2,6-dimethylpyridine, 2,4-dimethylpyridine and pyridine. A mutant with lost ability to degrade 2,6-dimethylpyridine was isolated during the growth of strain KM-4 rifR at 42 degrees C. Electrophoretic analysis of the plasmid from temperature sensitive mutant revealed the deletion the size of 26 Md from pBS320 plasmid.
Sigma-Aldrich
2,6-Lutidine, ReagentPlus®, 98%
Sigma-Aldrich
2,6-Lutidine
Supelco
2,6-Lutidine, analytical standard
Sigma-Aldrich
2,4-Lutidine, 99%