Merck
  • From 1D strands to extended molecular assemblies in the binary compounds of dithiooxamide and dithiobiurea with crown ethers.
From 1D strands to extended molecular assemblies in the binary compounds of dithiooxamide and dithiobiurea with crown ethers.
Organic & biomolecular chemistry (2003-09-13)
Yurii A Simonov, Marina S Fonari, Michael J Zaworotko, Heba Abourahma, Janusz Lipkowski, Edward V Ganin, Arkadii A Yavolovskii
ABSTRACT
The hydrogen-bonding networks for seven new binary compounds of dithiooxamide, (NH2CS)2 (dtox) and dithiobiurea (NH2CSNH)2 (dtur) with crown ethers, 18-crown-6 (18C6), 15-crown-5 (15C5), 12-crown-4 (12C4), cis-syn-cis-(DCHA), and cis-anti-cis-(DCHB) isomers of dicyclohexyl-18 -crown-6 are discussed. (15C5.dtox), (18C6.dtur) and (DCHB.dtur) afford one-dimensional hydrogen-bonded polymeric arrays where the components alternate. In (DCHA.2dtox) and (DCHB.2dtox) the similar hydrogen-bonded chains are further interlinked via dtox molecules to generate layered motifs. In (15C5.2dtur) the dtur molecules are self-assembled into layers via N-H...S hydrogen bonds. 15C5 spacers link adjacent layers into a three-dimensional network. In (12C4.dtur) the dtur molecules are arranged in chains. These chains alternate with the crown molecules attached to them through N-H...O hydrogen bonds in such a way that each 12C4 appears to be linked with four dtur molecules and vice versa thus providing a three-dimensional grid.
Sigma-Aldrich
Dithiooxamide, 97%