Merck
  • [The comparisons of fluorescence quenching between perylene and pyrene].
[The comparisons of fluorescence quenching between perylene and pyrene].
Guang pu xue yu guang pu fen xi = Guang pu (2003-09-02)
Z Chen, Z Zhang, X Yuan, G Zhang, F Bai
ABSTRACT
The fluorescence quenching of perylene and pyrene by dimethyl terephthalate (DMTP) and N,N-dimethylaniline (DMA) have been investigated. The results show that pyrene can form exciplex with DMTP or DMA at room temperature and perylene can only form exciplex with DMA. The fluorescence quenching date is in conformity with the Stern-Volmer equation: F0/F = 1 + KSV[Q] = 1 + Kq.tau 0[Q], the F0/F-[Q] straight lines are drawn. The Stren-Volmer constants and fluorescence quenching rate constants (KSV and Kq) are obtained. The Stern-Volmer quenching constants of pyrene is larger than that of perylene. The differences is due to the molecular structure of perylene which is not a typical large conjugated system and less coplanar configuration than pyrene.
Sigma-Aldrich
Dimethyl terephthalate, purum, ≥99.0% (GC)
Sigma-Aldrich
Dimethyl terephthalate, ReagentPlus®, ≥99%
Supelco
Dimethyl terephthalate, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland