Merck
  • The biodegradation of 1,3-dichlorobenzene by an adapted strain Bacillus cereus PF-11 derived from town-gas industrial effluent.
The biodegradation of 1,3-dichlorobenzene by an adapted strain Bacillus cereus PF-11 derived from town-gas industrial effluent.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering (2003-08-28)
L Wang, Q Zhou, B S Zhang, Z L Li, H Chua, D M Ren
ABSTRACT
In the present study, an adapted bacterium PF-11 with high 1,3-dichlorobenzene degradation capability was isolated from town-gas industrial effluent through continuous introducing of N-methyl-N'-nitro-N-nitrosoquanidine (NTG). In suitable condition, a degradation rate of 32 mg L(-1) d(-1) of 1,3-dichlorobenzene was obtained by strain PF-11 with effective chlorion release. Strain PF-11 was tentatively identified as gram-positive Bacillus cereus. The substrate specificity of the strain PF-11 was relatively low, and the degradation rate for different chlorobenzenes was in the order of monochlorobenzene > 1,3-dichlorobenzene > 1,2-dichlorobenzene. Initial oxidation step was molecular oxygen attacking chlorobenzene ring catalyzed by dioxygenase.
Sigma-Aldrich
1,3-Dichlorobenzene, puriss., ≥99.0% (GC)
Sigma-Aldrich
1,3-Dichlorobenzene, 98%