Merck
  • Parkinsonian mimetics induce aspects of unfolded protein response in death of dopaminergic neurons.
Parkinsonian mimetics induce aspects of unfolded protein response in death of dopaminergic neurons.
The Journal of biological chemistry (2003-02-25)
William Andrew Holtz, Karen Laurel O'Malley
ABSTRACT
Genes associated with Parkinson's disease (PD) have suggested a role for ubiquitin-proteasome dysfunction and aberrant protein degradation in this disorder. Inasmuch as oxidative stress has also been implicated in PD, the present study examined transcriptional changes mediated by the Parkinsonism-inducing neurotoxins 6-hydroxydopamine (6-OHDA) and 1-methyl-4-phenylpyridinium (MPP+) in a dopaminergic cell line. Microarray analysis of RNA isolated from toxin treated samples revealed that the stress-induced transcription factor CHOP/Gadd153 was dramatically up-regulated by both 6-OHDA and MPP+. Treatment with 6-OHDA also induced a large number of genes involved in endoplasmic reticulum stress and unfolded protein response (UPR) such as ER chaperones and elements of the ubiquitin-proteasome system. Reverse transcription-PCR, Western blotting, and immunocytochemical approaches were used to quantify and temporally order the UPR pathways involved in neurotoxin-induced cell death. 6-OHDA, but not MPP+, significantly increased hallmarks of UPR such as BiP, c-Jun, and processed Xbp1 mRNA. Both toxins increased the phosphorylation of UPR proteins, PERK and eIF2 alpha, but only 6-OHDA increased phosphorylation of c-Jun. Thus, 6-OHDA is capable of triggering multiple pathways associated with UPR, whereas MPP+ exhibits a more restricted response. The involvement of UPR in these widely used neurotoxin models supports the role of ubiquitin-proteasome pathway dysfunction in PD.
Sigma-Aldrich
Phosphorus(V) oxychloride, ReagentPlus®, 99%
Sigma-Aldrich
Phosphorus(V) oxychloride, 99.999%