Merck
Fern constituents: triterpenoids from Adiantum capillus-veneris.
Chemical & pharmaceutical bulletin (2002-09-19)
Takahisa Nakane, Yoshiko Maeda, Hideharu Ebihara, Yoko Arai, Kazuo Masuda, Akihito Takano, Hiroyuki Ageta, Kenji Shiojima, Shao-Qing Cai, Osama Bashir Abdel-Halim
ABSTRACT
Two new migrated hopane triterpenoids, viz. 4alpha-hydroxyfilican-3-one and fern-9(11)-en-12beta-ol, and olean-18-en-3-one and olean-12-en-3-one as the first example of oleanane compounds from Adiantum ferns were isolated along with many other known triterpenoids from Adiantum capillus-veneris of China and Egypt. Their structures were elucidated by spectroscopic analyses.
Sigma-Aldrich
Lithium aluminum hydride, pellets, reagent grade, 95%
Sigma-Aldrich
Lithium aluminum hydride, hydrogen-storage grade
Sigma-Aldrich
Lithium aluminum hydride solution, 2.3 M in 2-methyltetrahydrofuran
Sigma-Aldrich
Lithium aluminum hydride, powder, reagent grade, 95%
Sigma-Aldrich
Lithium aluminum hydride, ≥97.0% (gas-volumetric)
Sigma-Aldrich
Lithium aluminum hydride, ≥97.0% (gas-volumetric), tablet (5 g each)
Sigma-Aldrich
Lithium aluminum hydride solution, 0.5 M in 2-methoxyethyl ether
Sigma-Aldrich
Lithium aluminum hydride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Lithium aluminum hydride solution, 1.0 M in THF
Sigma-Aldrich
Lithium aluminum hydride solution, 2.0 M in THF