Merck
  • Background odour induces adaptation and sensitization of olfactory receptors in the antennae of houseflies.
Background odour induces adaptation and sensitization of olfactory receptors in the antennae of houseflies.
Medical and veterinary entomology (2002-07-12)
F J Kelling, F Ialenti, C J Den Otter
ABSTRACT
The presence of background odour was found to have a small but significant effect on the sensitivity of the antennal olfactory system of houseflies, Musca domestica Linnaeus (Diptera: Muscidae), to new pulses of odour. We show that cross-adaptation and cross-sensitization between a background odour of (+/-)-1-octen-3-ol and pulses of (+/-)-1-octen-3-ol, 2-pentanone and R-(+)-limonene can occur, confirming that olfactory receptor cells are sensitive to different odours. Background odour can increase the responses to low concentration odour pulses and decrease the responses to higher concentration odour pulses. It is suggested that background odour has a larger effect on olfactory receptor cells that respond with a tonic increase of spike frequency, giving information about the level of odour concentration, i.e. the 'static' environment. Cells that respond in a phasic way only provide information on the dynamics of the olfactory environment.
Product Description
Supelco
2-Pentanone, analytical standard
Sigma-Aldrich
2-Pentanone, ≥98%, FCC, FG
Sigma-Aldrich
2-Pentanone, suitable for HPLC, 99.5%
Sigma-Aldrich
2-Pentanone, ultrapure grade, ≥99.5%
Sigma-Aldrich
2-Pentanone, reagent grade, ≥90%