Merck
  • Dimerization of organic free radicals in solution. 1. Temperature dependent measurements.
Dimerization of organic free radicals in solution. 1. Temperature dependent measurements.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2002-05-08)
G Grampp, S Landgraf, K Rasmussen, S Strauss
ABSTRACT
Dimerization enthalpies and equilibrium constants have been determined for the radical anion of 2,3-dichloro-5,6-dicyano-p-benzoquinone (DDQ), the radical cations of N,N,N',N'-tetramethyl-p-phenylenediamine, N,N-dimethyl-p-phenylenediamine, 2,3,5,6-tetramethyl-p-phenylenediamine, N,N,N',N'-tetraethyl-p-phenylenediamine, N,N-diethyl-p-phenylenediamine and N,N,N'-trimethyl-p-phenylenediamine. Neutral radicals investigated are 2,2,6,6-tetramethylpiperidine-1-oxyl (TEMPO) and galvinoxyl. Solvent used was acetone, EtOH/Et2O-mixture (2:1 volume), propionitrile/butyronitrile-mixture (1:1 M) and dichloromethane. Measured dimerization enthalpies deltaHdim vary from -72.1 to -16.6 k/mol.
Sigma-Aldrich
Butyronitrile, purum, ≥99.0% (GC)
Sigma-Aldrich
Butyronitrile, ≥99%
Sigma-Aldrich
Galvinoxyl, free radical