Merck
  • The gas-phase on-line production of vanadium oxytrihalides, VOX3 and their identification by infrared spectroscopy.
The gas-phase on-line production of vanadium oxytrihalides, VOX3 and their identification by infrared spectroscopy.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2001-01-06)
M D Zidan, A W Allaf
ABSTRACT
A new route has been devised, leading to the production of VOX3 molecules where X=F, Br and I by an on-line process using vanadium oxytrichloride, VOCl3 as a starting compound passed over the following heated salts NaF, KBr and KI at 375, 700 and 550 degrees C, respectively. The products have been characterized by the IR spectra of their vapors. The low resolution gas phase on-line Fourier transform infrared spectra reported for the first time show strong bands with PQR type structure, centered at 1058, 1035, 1030 and 1025 cm(-1) assigned to the v1(a1), the O=V stretching fundamental mode of VOF3, VOCl3 VOBr3 and VOI3, respectively.
Sigma-Aldrich
Vanadium(V) oxychloride, 99%