Merck
  • Caffeic acid oligomers in Lithospermum erythrorhizon cell suspension cultures.
Caffeic acid oligomers in Lithospermum erythrorhizon cell suspension cultures.
Phytochemistry (2000-04-04)
H Yamamoto, K Inoue, K Yazaki
ABSTRACT
Lithospermum erythrorhizon cells cultured in pigment production (M-9) medium produced lithospermic acid B, a dimerized caffeic acid ester derivative, in quantities similar to the production of shikonin. The cells also produced a related dimer, (+)-rabdosiin. In Linsmaier-Skoog liquid medium, which suppresses shikonin production, both lithospermic acid B and (+)-rabdosiin were still formed. Lithospermic acid, a caffeic acid-rosmarinic acid conjugate, was isolated as a main constituent in Lithospermum hairy root cultures. In the aerial parts of L. erythrorhizon, the content of these phenylpropanoid oligomers was relatively low compared to that of rosmarinic acid.
Product Description
Supelco
Salvianolic acid B, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Salvianolic acid B, ≥94% (HPLC)