Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • Long-term effects of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate and 6-nitro-7-sulphamoylbenzo(f)quinoxaline-2,3-dione in the rat basal ganglia: calcification, changes in glutamate receptors and glial reactions.
Long-term effects of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate and 6-nitro-7-sulphamoylbenzo(f)quinoxaline-2,3-dione in the rat basal ganglia: calcification, changes in glutamate receptors and glial reactions.
Neuroscience (1999-12-29)
V Petegnief, J Saura, D Dewar, D J Cummins, M Dragunow, N Mahy
ABSTRACT
Previous data from our laboratory indicate that 25 mM ibotenic acid induces intracellular calcifications in the rat basal forebrain. Because of the lack of specificity of ibotenic acid for a glutamate receptor subtype, a dose-response study with alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate was undertaken and calcified areas (identified with Alizarin Red staining) as well as astro- and microglial reactions (by autoradiography with [3H]lazabemide and [3H]Ro 5-4864) were quantified at one month post-lesion. alpha-Amino-3-hydroxy-5-methyl-4-isoxazole propionate administered into the globus pallidus induced, in a dose-dependent manner, the formation of calcium deposits and the activation of both glial cells, the microglial reaction being particularly robust. From this study, a dose of 5.4 mM alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate was selected for further experiments. [3H]alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate, [3H]dizocilpine maleate and [3H]PN 200-110 binding in vitro were performed to assess autoradiographically whether the tissue damage was associated with changes in glutamate receptors and calcium channel binding sites. In the alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate-treated animals, the specific binding of [3H]alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate was significantly reduced by 28% in the lesioned ventral pallidum, whereas it was unchanged in the globus pallidus and substantia innominata. In these three nuclei, calcifications developed and an increase in both glial markers was measured. In contrast, the binding of [3H]PN 200-110 and [3H]dizocilpine maleate were unaffected. Co-injection of 15 mM 6-nitro-7-sulphamoylbenzo(f)quinoxaline-2,3-dione, a selective alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate/kainate receptor antagonist, prevented the formation of calcium concretions, the microglial reaction and the decrease in [3H]alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate binding but it failed to inhibit totally the astroglial reaction induced by alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate. This may suggest that the microglial reaction and calcification take place through different mechanisms from the astrogliosis associated with the neuronal loss. In conclusion, acute administration of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate in the rat globus pallidus elicits a dose-dependent calcification process associated with a chronic reaction of astrocytes and microglia. alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate-induced injury is accompanied by a slight reduction of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate receptors in the ventral pallidum, whereas the binding of N-methyl-D-aspartate and L-type calcium channels receptors remains unchanged in any lesioned nucleus.
Sigma-Aldrich
Lazabemide hydrate, ≥97% (HPLC)