Merck
  • Intracellular pH on adrenergic-stimulated cAMP and cGMP production in rat pinealocytes.
Intracellular pH on adrenergic-stimulated cAMP and cGMP production in rat pinealocytes.
The American journal of physiology (1991-10-01)
A K Ho, M Girard, I Young, C L Chik
ABSTRACT
In rat pinealocytes, activation of alpha 1-adrenergic receptors leads to increases in intracellular pH (pHi). In this study, the role of pHi on adrenergic regulation of cyclic nucleotide accumulation was investigated using ammonium chloride, which increased pHi, and sodium propionate, which reduced pHi. Ammonium chloride significantly enhanced the norepinephrine-stimulated adenosine 3',5'-cyclic monophosphate (cAMP) and guanosine 3',5'-cyclic monophosphate (cGMP) responses, while it selectively potentiated the isoproterenol (ISO)-stimulated cGMP response. Reduction of pHi by sodium propionate reduced the norepinephrine-stimulated cGMP accumulation by 70%, and its effect on the ISO-stimulated cGMP response was stimulatory. Treatment with sodium propionate effectively neutralized the enhancing effects of ammonium chloride on the adrenergic-stimulated cAMP and cGMP responses. These effects of sodium propionate and ammonium chloride on cyclic nucleotides appeared to reflect altered rate of synthesis, and they were also in part secondary to changes in intracellular Ca2+. Our findings indicate that the receptor-mediated changes in pHi may play an integral part in the adrenergic regulation of cAMP and cGMP production in rat pinealocytes.
Sigma-Aldrich
2′,7′-Bis(2-carboxyethyl)-5(6)-carboxyfluorescein acetoxymethyl ester, 1 mg/mL in DMSO