Merck
  • Inhibition of H3K9 methyltransferases G9a/GLP prevents ototoxicity and ongoing hair cell death.
Inhibition of H3K9 methyltransferases G9a/GLP prevents ototoxicity and ongoing hair cell death.
Cell death & disease (2013-02-23)
H Yu, Q Lin, Y Wang, Y He, S Fu, H Jiang, Y Yu, S Sun, Y Chen, J Shou, H Li
ABSTRACT
Sensorineural hearing loss (SNHL) is one of the most common sensory defects in humans. Hair cells are vulnerable to various ototoxic insults. Effective prevention of hair cell loss remains an unmet medical need. Apoptotic hair cell death, which involves active regulation of transcription, accounts for the majority of aminoglycoside-induced hair cells loss. As one of the important epigenetic covalent modifications, histone methylation is involved in the regulation of gene expression, development and reaction to injury. In particular, H3K9 dimethylation (H3K9me2) is critical for euchromatin gene silencing. In the present study, we examined the roles of two highly homologous histone methyltransfereases responsible for this modification, G9a/G9a-like protein (GLP), in the reaction to aminoglycoside-induced hair cell damage. We observed a rapid increase of H3K9me2 upon hair cell damage in organotypic cochlear cultures. Treatment with the G9a/GLP-specific inhibitors, BIX01294 or UNC0638, reduced the level of H3K9me2 and prevented hair cells from death. Local delivery of BIX01294 also prevented neomycin-induced in vivo auditory hair cell loss in the organ of Corti in a mouse damage model. It is unlikely that BIX01294 functions through blocking aminoglycoside absorption as it does not interfere with aminoglycoside uptaking by hair cells in the organotypic cochlear cultures. Our data revealed a novel role of histone methylation in otoprotection, which is of potential therapeutic value for SNHL management.
Product Description
Supelco
Neomycin trisulfate salt hydrate, VETRANAL®, analytical standard
Sigma-Aldrich
Neomycin trisulfate salt hydrate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
UNC0638 hydrate, ≥98% (HPLC)
Sigma-Aldrich
Neomycin trisulfate salt hydrate, meets USP testing specifications, powder
Sigma-Aldrich
Neomycin trisulfate salt hydrate, powder
Sigma-Aldrich
Neomycin trisulfate salt hydrate, suitable for plant cell culture
Neomycin sulphate, European Pharmacopoeia (EP) Reference Standard