Merck
  • Copy number variation and selection during reprogramming to pluripotency.
Copy number variation and selection during reprogramming to pluripotency.
Nature (2011-03-04)
Samer M Hussein, Nizar N Batada, Sanna Vuoristo, Reagan W Ching, Reija Autio, Elisa Närvä, Siemon Ng, Michel Sourour, Riikka Hämäläinen, Cia Olsson, Karolina Lundin, Milla Mikkola, Ras Trokovic, Michael Peitz, Oliver Brüstle, David P Bazett-Jones, Kari Alitalo, Riitta Lahesmaa, Andras Nagy, Timo Otonkoski
ABSTRACT
The mechanisms underlying the low efficiency of reprogramming somatic cells into induced pluripotent stem (iPS) cells are poorly understood. There is a clear need to study whether the reprogramming process itself compromises genomic integrity and, through this, the efficiency of iPS cell establishment. Using a high-resolution single nucleotide polymorphism array, we compared copy number variations (CNVs) of different passages of human iPS cells with their fibroblast cell origins and with human embryonic stem (ES) cells. Here we show that significantly more CNVs are present in early-passage human iPS cells than intermediate passage human iPS cells, fibroblasts or human ES cells. Most CNVs are formed de novo and generate genetic mosaicism in early-passage human iPS cells. Most of these novel CNVs rendered the affected cells at a selective disadvantage. Remarkably, expansion of human iPS cells in culture selects rapidly against mutated cells, driving the lines towards a genetic state resembling human ES cells.
Sigma-Aldrich
LuminoCt® qPCR ReadyMix, For fast probe-based quantitative PCR