Merck
  • Some characteristics of a new glycopeptidase acting on aspartylglycosylamine linkages.
Some characteristics of a new glycopeptidase acting on aspartylglycosylamine linkages.
Journal of biochemistry (1978-12-01)
N Takahashi, H Nishibe
PMID738997
ABSTRACT
A new type of glycopeptidase hydrolyzing beta-aspartylglycosylamine linkages was partially purified from almond emulsin by chromatography on Sephadex G-200 and DE 52. The enzyme degraded stem bromelain glycopeptide, Asn-Asn(Man3,Xyl1,Fuc1,GlcNAc2)-Glu-Ser-Ser, to yield equimolar amounts of intact oligosaccharide, peptide (Asn-Asp-Glu-Ser-Ser), and ammonia. The Km value for the stem bromelain glycopeptide was 4 mM, and the optimum pH was 5.2. The enzyme was markedly inhibited by 10 mM Cu2+, Fe3+, and Zn2+. Thiol inhibitors and actinomycete protease inhibitors had no effect. The glycopeptides used as substrates were prepared from stem bromelain, ovalbumin or ovotransferrin. The enzyme hydrolyzed glycopeptides with 3-11 amino acid residues, whereas it did not hydrolyze glycopeptides with 1-2 amino acid residues. Furthermore, Asn-oligosaccharide was not inhibitory to the enzyme.
Sigma-Aldrich
PNGase F from Elizabethkingia meningoseptica, BioReagent, ≥95% (SDS-PAGE), for proteomics
Sigma-Aldrich
Glycopeptidase A from almonds, buffered aqueous glycerol solution, ≥0.05 unit/mL
Sigma-Aldrich
PNGase F from Elizabethkingia miricola, buffered aqueous solution
Sigma-Aldrich
PNGase F from Elizabethkingia meningoseptica, lyophilized powder, recombinant, expressed in E. coli
Sigma-Aldrich
PNGase F from Elizabethkingia meningoseptica, ready-to-use solution, recombinant, expressed in E. coli
Sigma-Aldrich
PNGase F from Elizabethkingia meningoseptica, recombinant, expressed in E. coli, set of 100 units nanomolar unit