Merck
Isolation of a functional human gene for brain creatine kinase.
The Journal of biological chemistry (1988-02-15)
G H Daouk, R Kaddurah-Daouk, S Putney, R Kingston, P Schimmel
ABSTRACT
There is evidence that the gene for the B isozyme of creatine kinase is regulated during cell differentiation, is under hormonal control, and is activated in a small cell lung carcinoma. In order to investigate further the mechanisms of these processes, the human gene was isolated and the structure of the promoter region was determined. A human DNA fragment of 8 kilobase pairs was shown to encompass the entire coding region and 850 base pairs (bp) of the 5'-flanking sequence. This fragment was transfected into three cell lines and shown to express functional enzyme. The 5'-end of the gene is split by a 230-bp intron that is located 12 bp upstream of the initiator ATG codon. Transcription initiation occurs at a site that is approximately 69 bp upstream of the 5'-end of this intron. The DNA sequence in the region upstream of the 5'-end of the mRNA is suggestive of two superimposed promoters that contain additional sequence elements that are known to regulate expression of other eukaryote genes. The 5'-region also has a remarkable homology to the overlapping promoters of the adenovirus EIIaE gene. These elements collectively form the basis for initial investigations of how this gene is controlled.
Sigma-Aldrich
Creatine Phosphokinase from bovine heart, Type III, salt-free, lyophilized powder, ≥30 units/mg protein
Sigma-Aldrich
Creatine Phosphokinase from rabbit muscle, Type I, salt-free, lyophilized powder, ≥150 units/mg protein