Merck
  • CBP/p300 and SIRT1 are involved in transcriptional regulation of S-phase specific histone genes.
CBP/p300 and SIRT1 are involved in transcriptional regulation of S-phase specific histone genes.
PloS one (2011-07-27)
Hongpeng He, Fa-Xing Yu, Chi Sun, Yan Luo
ABSTRACT
Histones constitute a type of essential nuclear proteins important for chromatin structure and functions. The expression of major histones is strictly confined to the S phase of a cell cycle and tightly coupled to DNA replication. With RT-qPCR and ChIP assays, we investigated transcriptional regulation of the S-phase specific histone genes and found that the acetylation level of histones on core histone gene promoters fluctuated during cell cycle in a pattern similar to RNA polymerase II association. Further, we showed that CBP/p300 and SIRT1 were recruited to histone gene promoters in an NPAT-dependent manner, knockdown of which affected histone acetylation on histone gene promoters and histone gene transcription. These observations contribute to further understanding of the mechanism by which the expression of canonical histone genes is regulated, and also implicate a link between histone expression and DNA damage repair and cell metabolism.
Sigma-Aldrich
Formaldehyde solution, 36.5-38% in H2O
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Anti-acetyl-Histone H4 Antibody, serum, Upstate®
Sigma-Aldrich
Anti-acetyl-Histone H4 Antibody, 1 mg/mL, Upstate®
Sigma-Aldrich
Formaldehyde solution, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Supelco
Resveratrol, analytical standard
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)
Supelco
Formaldehyde in Water
Supelco
Resveratrol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Anti-acetyl-Histone H3 Antibody, from rabbit
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse