Merck
  • A common variant in PNPLA3, which encodes adiponutrin, is associated with liver fat content in humans.
A common variant in PNPLA3, which encodes adiponutrin, is associated with liver fat content in humans.
Diabetologia (2009-02-19)
A Kotronen, L E Johansson, L M Johansson, C Roos, J Westerbacka, A Hamsten, R Bergholm, P Arkkila, J Arola, T Kiviluoto, R M Fisher, E Ehrenborg, M Orho-Melander, M Ridderstråle, L Groop, H Yki-Järvinen
ABSTRACT
It has recently been suggested that the rs738409 G allele in PNPLA3, which encodes adiponutrin, is strongly associated with increased liver fat content in three different ethnic groups. The aims of the present study were as follows: (1) to try to replicate these findings in European individuals with quantitative measures of hepatic fat content; (2) to study whether the polymorphism influences hepatic and adipose tissue insulin sensitivity; and (3) to investigate whether PNPLA3 expression is altered in the human fatty liver. We genotyped 291 Finnish individuals in whom liver fat had been measured using proton magnetic resonance spectroscopy. Hepatic PNPLA3 expression was measured in 32 participants. Hepatic and adipose tissue insulin sensitivities were measured using a euglycaemic-hyperinsulinaemic (insulin infusion 0.3 mU kg(-1) min(-1)) clamp technique combined with infusion of [3-(3)H]glucose in 109 participants. The rs738409 G allele in PNPLA3 was associated with increased quantitative measures of liver fat content (p = 0.011) and serum aspartate aminotransferase concentrations (p = 0.002) independently of age, sex and BMI. Fasting serum insulin and hepatic and adipose tissue insulin sensitivity were related to liver fat content independently of genotype status. PNPLA3 mRNA expression in the liver was positively related to obesity (r = 0.62, p < 0.0001) and to liver fat content (r = 0.58, p = 0.025) in participants who were not morbidly obese (BMI < 40 kg/m(2)). A common variant in PNPLA3 increases the risk of hepatic steatosis in humans.
Sigma-Aldrich
Lipase from Candida sp., recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
Sigma-Aldrich
Lipase from Rhizopus niveus, powder (fine), ≥1.5 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
Sigma-Aldrich
Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg
Sigma-Aldrich
Lipase from Aspergillus oryzae, ≥20,000 U/g
Sigma-Aldrich
Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
Sigma-Aldrich
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid
Sigma-Aldrich
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg solid
Sigma-Aldrich
Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
Sigma-Aldrich
Lipase from Pseudomonas cepacia, powder, light beige, ≥30 U/mg
Sigma-Aldrich
Lipase A Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~2 U/mg
Sigma-Aldrich
Lipase immobilized from Candida antarctica, beads, slightly brown, >2 U/mg
Sigma-Aldrich
Lipase from Aspergillus niger, powder (fine), ~200 U/g
Sigma-Aldrich
Lipase from Mucor javanicus, lyophilized powder, ≥300 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase B Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~9 U/mg