Merck
  • Identification, cloning, expression, and purification of three novel human calcium-independent phospholipase A2 family members possessing triacylglycerol lipase and acylglycerol transacylase activities.
Identification, cloning, expression, and purification of three novel human calcium-independent phospholipase A2 family members possessing triacylglycerol lipase and acylglycerol transacylase activities.
The Journal of biological chemistry (2004-09-15)
Christopher M Jenkins, David J Mancuso, Wei Yan, Harold F Sims, Beverly Gibson, Richard W Gross
ABSTRACT
Genetic knockout of hormone-sensitive lipase in mice has implicated the presence of other intracellular triacylglycerol (TAG) lipases mediating TAG hydrolysis in adipocytes. Despite intense interest in these TAG lipases, their molecular identities thus far are largely unknown. Sequence data base searches for proteins containing calcium-independent phospholipase A2 (iPLA2) dual signature nucleotide ((G/A)XGXXG) and lipase (GXSXG) consensus sequence motifs identified a novel subfamily of three putative iPLA2/lipase family members designated iPLA2epsilon, iPLA2zeta, and iPLA2eta (previously named adiponutrin, TTS-2.2, and GS2, respectively) of previously unknown catalytic function. Herein we describe the cloning, heterologous expression, and affinity purification of the three human isoforms of this iPLA2 subfamily in Sf9 cells, and we demonstrate that each possesses abundant TAG lipase activity. Moreover, iPLA2epsilon, iPLA2zeta, and iPLA2eta also possess acylglycerol transacylase activity utilizing mono-olein as an acyl donor which, in the presence of mono-olein or diolein acceptors, results in the synthesis of diolein and triolein, respectively. (E)-6-(Bromomethylene)-3-(1-naphthalenyl)-2H-tetrahydropyran-2-one, a mechanism-based suicide substrate inhibitor of all known iPLA2s, inhibits the triglyceride lipase activity of each of the three isoforms similarly (IC50=0.1-0.5 microm). Quantitative PCR revealed dramatically increased expression of iPLA2epsilon and iPLA2zeta transcripts during the hormone-induced differentiation of 3T3-L1 cells into adipocytes and identified the presence of all three iPLA2 isoforms in human SW872 liposarcoma cells. Collectively, these results identify three novel TAG lipases/acylglycerol transacylases that likely participate in TAG hydrolysis and the acyl-CoA independent transacylation of acylglycerols, thereby facilitating energy mobilization and storage in adipocytes.
Sigma-Aldrich
Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
Sigma-Aldrich
Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
Sigma-Aldrich
Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
Sigma-Aldrich
Lipase from Rhizopus niveus, powder (fine), ≥1.5 U/mg
Sigma-Aldrich
Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid
Sigma-Aldrich
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg solid
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
Sigma-Aldrich
Lipase from Aspergillus oryzae, ≥20,000 U/g
Sigma-Aldrich
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Candida sp., recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Pseudomonas cepacia, powder, light beige, ≥30 U/mg
Sigma-Aldrich
Lipase from Aspergillus niger, powder (fine), ~200 U/g
Sigma-Aldrich
Lipase A Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~2 U/mg
Sigma-Aldrich
Lipase B Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~9 U/mg
Sigma-Aldrich
Lipase from Mucor javanicus, lyophilized powder, ≥300 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase immobilized from Candida antarctica, beads, slightly brown, >2 U/mg