Merck
Hydrolysis of retinyl esters by pancreatic triglyceride lipase.
Biochemistry (2000-04-19)
A M van Bennekum, E A Fisher, W S Blaner, E H Harrison
ABSTRACT
Previously [van Bennekum, A. M., et al. (1999) Biochemistry 38, 4150-4156] we showed that carboxyl ester lipase (CEL)-deficient (CELKO) mice have normal levels of pancreatic, bile salt-dependent retinyl ester hydrolase (REH) activity. In the present study, we further investigated this non-CEL REH activity in pancreas homogenates of CELKO and wild-type (WT) mice, and rats. REH activity was detected in both the presence and absence of tri- and dihydroxy bile salts in rats, WT mice, and CELKO mice. In contrast, pancreatic cholesteryl ester hydrolase (CEH) activity was only detected in the presence of trihydroxy bile salts and only in rats and WT mice, consistent with CEL-mediated cholesteryl ester hydrolysis. Enzyme assays of pancreatic triglyceride lipase (PTL) showed that there was a colipase-stimulated REH activity in rat and mouse (WT and CELKO) pancreas, consistent with hydrolysis of retinyl ester (RE) by PTL. Pancreatic enzyme activities related to either CEL or PTL were separated using DEAE-chromatography. In both rats and mice (WT and CELKO), REH activity could be attributed mainly to PTL, and to a much smaller extent to CEL. Finally, purified human PTL exhibited similar enzymatic characteristics for triglyceride hydrolysis as well as for retinyl ester hydrolysis, indicating that RE is a substrate for PTL in vivo. Altogether, these studies clearly show that PTL is the major pancreatic REH activity in mice, as well as in rats.
Sigma-Aldrich
Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
Sigma-Aldrich
Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
Sigma-Aldrich
Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
Sigma-Aldrich
Lipase from Rhizopus niveus, powder (fine), ≥1.5 U/mg
Sigma-Aldrich
Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid
Sigma-Aldrich
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg solid
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
Sigma-Aldrich
Lipase from Aspergillus oryzae, ≥20,000 U/g
Sigma-Aldrich
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Candida sp., recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Pseudomonas cepacia, powder, light beige, ≥30 U/mg
Sigma-Aldrich
Lipase from Aspergillus niger, powder (fine), ~200 U/g
Sigma-Aldrich
Lipase A Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~2 U/mg
Sigma-Aldrich
Lipase B Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~9 U/mg
Sigma-Aldrich
Lipase from Mucor javanicus, lyophilized powder, ≥300 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase immobilized from Candida antarctica, beads, slightly brown, >2 U/mg