Merck
  • Biological activity, membrane-targeting modification, and crystallization of soluble human decay accelerating factor expressed in E. coli.
Biological activity, membrane-targeting modification, and crystallization of soluble human decay accelerating factor expressed in E. coli.
Protein science : a publication of the Protein Society (2004-08-24)
Jennifer White, Petra Lukacik, Dirk Esser, Michael Steward, Naomi Giddings, Jeremy R Bright, Sarah J Fritchley, B Paul Morgan, Susan M Lea, Geoffrey P Smith, Richard A G Smith
ABSTRACT
Decay-accelerating factor (DAF, CD55) is a glycophosphatidyl inositol-anchored glycoprotein that regulates the activity of C3 and C5 convertases. In addition to understanding the mechanism of complement inhibition by DAF through structural studies, there is also an interest in the possible therapeutic potential of the molecule. In this report we describe the cloning, expression in Escherichia coli, isolation and membrane-targeting modification of the four short consensus repeat domains of soluble human DAF with an additional C-terminal cysteine residue to permit site-specific modification. The purified refolded recombinant protein was active against both classical and alternative pathway assays of complement activation and had similar biological activity to soluble human DAF expressed in Pichia pastoris. Modification with a membrane-localizing peptide restored cell binding and gave a large increase in antihemolytic potency. These data suggested that the recombinant DAF was correctly folded and suitable for structural studies as well as being the basis for a DAF-derived therapeutic. Crystals of the E. coli-derived protein were obtained and diffracted to 2.2 A, thus permitting the first detailed X-ray crystallography studies on a functionally active human complement regulator protein with direct therapeutic potential.