Merck
  • alpha-Glucosidase and alpha-amylase inhibitory activities of saponins from traditional Chinese medicines in the treatment of diabetes mellitus.
alpha-Glucosidase and alpha-amylase inhibitory activities of saponins from traditional Chinese medicines in the treatment of diabetes mellitus.
Die Pharmazie (2013-05-25)
Fang Dou, Miaomiao Xi, Junxian Wang, Xiangrong Tian, Liangjian Hong, Haifeng Tang, Aidong Wen
ABSTRACT
Extracts of eleven traditional Chinese medicines (TCM) with a reputation of usefulness in treating diabetes mellitus were examined for alpha-glucosidase and alpha-amylase inhibitory activities in vitro. The extract with the highest activity was selected for further characterization. The extract of the root bark of Aralia taibaiensis (EAT) outperformed other extracts in the assays with IC50 values of 0.48 +/- 0.01 mg/mL (BSG), 0.41 +/- 0.02mg/mL (SCG), 0.61 +/- .03mg/mL (BLA) and 0.77 +/- 0.03mg/mL (PPA), respectively. To identify which constituents were responsible for the activities, thirteen triterpenoid saponins were isolated from EAT and examined for their inhibitory effects against alpha-glucosidase and alpha-amylase. The results revealed that saponins 2, 3, 4 (IC50: 0.83 +/- 0.05 microM for BSG and IC50: 0.72 +/- 0.03 microM for SCG), 5, 6, 7, 9, 10, 11 and 12 (IC50: 1.07 +/- 0.04 micro.M for BSG and IC50: 0.93 +/- 0.05 micro.M for SCG) showed alpha-glucosidase inhibitory activities, while 2, 3, 4 (IC50: 0.93 +/- 0.04 micro.M for PPA), 5, 6, 7, 9, 10, 11 and 12 (IC50: 1.02 +/- 0.03 micro.M for PPA) possessed significant alpha-amylase inhibitory activities. In addition, the structure-activity relationship of the thirteen saponins was discussed based on their structure features and diabetes mellitus related activities. It is suggested that the glucuronic acid unit at C-3 of the aglycone is the imperative functional group of the antidiabetic activities, and two characteristic structural features are responsible for the remarkable alpha-glucosidase and alpha-amylase inhibitory activities.
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~380 U/mg
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~50 U/mg
Sigma-Aldrich
Taka-Diastase from Aspergillus oryzae, powder, slightly beige, ~100 U/mg
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, ≥400 units/mg protein (Lowry)
Sigma-Aldrich
α-Amylase from porcine pancreas, Type I-A, PMSF treated, saline suspension, 700-1400 units/mg protein (E1%/280)
Sigma-Aldrich
α-Amylase from Bacillus sp., Type II-A, lyophilized powder, ≥1,500 units/mg protein (biuret)
Sigma-Aldrich
α-Amylase, heat-stable, solution, for use in Total Dietary Fiber Assay, TDF-100A
Sigma-Aldrich
α-Amylase from porcine pancreas, PMSF Treated, Type I-A, saline suspension, ≥1000 units/mg protein (E1%/280)
Sigma-Aldrich
α-Amylase from Bacillus sp., liquid
Sigma-Aldrich
α-Glucosidase from Saccharomyces cerevisiae, recombinant, expressed in proprietary host, lyophilized powder, ≥100 units/mg protein
Sigma-Aldrich
α-Glucosidase from rice, Type V, ammonium sulfate suspension, 40-80 units/mg protein
Sigma-Aldrich
α-Glucosidase from Saccharomyces cerevisiae, Type I, lyophilized powder, ≥10 units/mg protein (using p-nitrophenyl α-D-glucoside as substrate.)
Sigma-Aldrich
α-Amylase from Bacillus licheniformis, lyophilized powder, 500-1,500 units/mg protein, 93-100% (SDS-PAGE)
Sigma-Aldrich
α-Amylase from Bacillus licheniformis, Type XII-A, saline solution, ≥500 units/mg protein (biuret)
Supelco
α-Amylase from Bacillus licheniformis, suitable for determination of starch (Kit STA-20)
Sigma-Aldrich
α-Amylase from human saliva, Type XIII-A, lyophilized powder, 300-1,500 units/mg protein
Sigma-Aldrich
α-Amylase from human saliva, Type IX-A, lyophilized powder, 1,000-3,000 units/mg protein
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, aqueous solution, ≥800 FAU/g
Sigma-Aldrich
α-Amylase from Bacillus amyloliquefaciens, liquid, ≥250 units/g
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, powder, ~30 U/mg
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, powder, ~1.5 U/mg (~0.2 U acc. to Willstätter)
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, ≥150 units/mg protein (biuret)
Sigma-Aldrich
α-Glucosidase from Bacillus stearothermophilus, lyophilized powder, ≥50 units/mg protein