Merck
  • Effects of a mono-component endo-xylanase supplementation on the nutritive value of wheat-based broiler diets.
Effects of a mono-component endo-xylanase supplementation on the nutritive value of wheat-based broiler diets.
British poultry science (2013-02-13)
M Francesch, A M Pérez-Vendrell, J Broz
ABSTRACT
1. Two trials were conducted to evaluate the effects of a mono-component thermostable endo-1,4-β-xylanase derived from Thermomyces lanuginosus on the nutritive value of wheat-based broiler diets. In a 5-week growth trial, the efficacy of xylanase supplementation at 0, 100, 150, 200, 400 and 4000 FXU/kg diet was evaluated. A short-term balance trial was carried out according to a 4 × 2 factorial arrangement of treatments, involving 4 wheat cultivars and endo-xylanase at 0 or 200 FXU/kg. 2. In the growth trial, enzyme supplementation from 0 to 400 FXU/kg reduced feed intake and improved feed conversion linearly. Digesta viscosity was significantly reduced by all enzyme inclusion levels by 49·6-56·9%, in a quadratic manner. 3. In the balance trial, xylanase supplementation resulted in a significant improvement of protein, lipid and dry matter apparent digestibility coefficients of diets, accompanied by improved dietary AME(N) values. There was a significant wheat × enzyme interaction on AME(N) and lipid digestibility. There was a significant effect of wheat cultivar on dry matter digestibility.
Sigma-Aldrich
Cellulase from Aspergillus niger, powder, ≥0.3 units/mg solid
Sigma-Aldrich
Cellulase from Aspergillus niger, powder, off-white, ~0.8 U/mg
Sigma-Aldrich
Cellulase from Trichoderma reesei, aqueous solution, ≥700 units/g
Sigma-Aldrich
Cellulase from Trichoderma sp., BioReagent, suitable for plant cell culture, 3-10 units/mg solid
Sigma-Aldrich
Cellulase from Trichoderma reesei ATCC 26921, lyophilized powder, ≥1 unit/mg solid
Sigma-Aldrich
Cellulase from Trichoderma sp., powder, ≥5,000 units/g solid