Merck
  • [Nutritional factors of inflammation induction or lipid mechanism of intestinal endotoxin transport].
[Nutritional factors of inflammation induction or lipid mechanism of intestinal endotoxin transport].
Fiziologiia cheloveka (2013-02-12)
P L Okorokov, I A Anikhovskaia, M M Iakovleva, S I Lazareva, A A Melamud, M V Meshkov, I M Salakhov, G G Énukidze, M Iu Iakovlev
ABSTRACT
Food behavior disorderes (high fat diet) and systematic strong drinks use could be the most important causes of excess intake of intestinal endotoxin (ET) in circulation (endotoxin aggression development) and induction of inflammation. In this work we investigated antiendotoxin immunity activity, ET level and some biochemical parameters in two group of patients, which were treated with orlistat or medical starvation. We suggest that lipid component of food takes a part in mechanism of ET delivery in bloodstream and its recycling in hydrophobic form. High serum ET level in obesity patients could be direct corollary of excess fat intake and strong drinks use. It provides increased lipid absorption in circulation. Fatty tissue could be depot of ET hydrophobic form in organism. We also suggest that lipid mechanism takes a part in ET deposition in bloodstream. Prolonged starvation and anorexia leads to development inflammatory changing like systematic inflammatory response syndrome (SIRS).
Sigma-Aldrich
Lipase from Candida sp., recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Aspergillus oryzae, ≥20,000 U/g
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
Sigma-Aldrich
Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
Sigma-Aldrich
Lipase from Rhizopus niveus, powder (fine), ≥1.5 U/mg
Sigma-Aldrich
Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
Sigma-Aldrich
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Aspergillus oryzae, solution, ≥100,000 U/g, white, beige
Sigma-Aldrich
Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
Sigma-Aldrich
Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg solid
Sigma-Aldrich
Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg
Sigma-Aldrich
Lipase B Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~9 U/mg
Sigma-Aldrich
Lipase from Aspergillus niger, powder (fine), ~200 U/g
Sigma-Aldrich
Lipase from Pseudomonas cepacia, powder, light beige, ≥30 U/mg
Sigma-Aldrich
Lipase immobilized from Candida antarctica, beads, slightly brown, >2 U/mg
Sigma-Aldrich
Lipase A Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~2 U/mg
Sigma-Aldrich
Lipase from Mucor javanicus, lyophilized powder, ≥300 units/mg solid (using olive oil)