Merck
  • Evaluation of lysozyme-HCl for the treatment of chalkbrood disease in honey bee colonies.
Evaluation of lysozyme-HCl for the treatment of chalkbrood disease in honey bee colonies.
Journal of economic entomology (2013-01-30)
A Van Haga, B A Keddie, S F Pernal
ABSTRACT
Chalkbrood, caused by Ascosphaera apis (Maassen and Claussen) Olive and Spiltor, is a cosmopolitan fungal disease of honey bee larvae (Apis mellifera L.) for which there is no chemotherapeutic control. We evaluated the efficacy of lysozyme-HCl, an inexpensive food-grade antimicrobial extracted from hen egg white, for the treatment of chalkbrood disease in honey bee colonies. Our study compared three doses of lysozyme-HCl in sugar syrup (600, 3,000, and 6,000 mg) administered weekly for 3 wk among chalkbrood-inoculated colonies, colonies that were inoculated but remained untreated, and colonies neither inoculated or treated. Lysozyme-HCl at the highest dose evaluated was found to suppress development of chalkbrood disease in inoculated colonies to levels observed in uninoculated, untreated colonies, and did not adversely affect adult bee survival or brood production. Honey production was significantly negatively correlated with increased disease severity but there were no significant differences in winter survival among treatment groups. Based on our results, lysozyme-HCl appears to be a promising, safe therapeutic agent for the control of chalkbrood in honey bee colonies.
Sigma-Aldrich
Lysozyme from human neutrophils, ≥95% (SDS-PAGE), lyophilized powder, ≥100,000 units/mg protein (E1%/280)
Sigma-Aldrich
Lysozyme human, Lysobac, recombinant, expressed in rice, lyophilized powder, ≥100,000 units/mg protein
Sigma-Aldrich
Lysozyme from chicken egg white, 10 mg/mL
Sigma-Aldrich
Lysozyme from chicken egg white, dialyzed, lyophilized, powder, ~100000 U/mg
Sigma-Aldrich
Lysozyme from chicken egg white, powder (crystalline), 70000-140000 U/mg
Sigma-Aldrich
Lysozyme from chicken egg white, aseptically filled
Sigma-Aldrich
Lysozyme from chicken egg white, powder or granules, ≥39,000 units/mg protein
Sigma-Aldrich
Lysozyme chloride form from chicken egg white, Grade VI, ≥35,000 units/mg protein (E1%/282)
Sigma-Aldrich
Lysozyme from chicken egg white, For use as a marker in SDS-PAGE
Sigma-Aldrich
Lysozyme from chicken egg white, BioUltra, lyophilized powder, ≥98% (SDS-PAGE), ≥40,000 units/mg protein