Merck
  • [Pharmacological correction of nonspecific resistance and production of proinflammatory cytokines during chronic intoxication with organophosphorus compound VX].
[Pharmacological correction of nonspecific resistance and production of proinflammatory cytokines during chronic intoxication with organophosphorus compound VX].
Eksperimental'naia i klinicheskaia farmakologiia (2013-01-18)
P F Zabrodskiĭ, V A Grishin
ABSTRACT
It is established in experiments on noninbred rats that the use of imunofan (20 mg/kg daily) and polyoxidonium (150 mg/kg daily) for 7 days on the background of chronic intoxication with organophosphorus agent VX (0.01 LD50, single daily treatment for 30 days) resulted in almost complete recovery of phagocytic-metabolic activity of neutrophils, the content of lysozyme, cationic protein of platelet, and levels of proinflammatory cytokines TNFa, IL-1b and IL-6 in the blood. The administration of T-activin (20 mg/kg daily for 7 days) restores these parameters insignificantly. The maximum overall stimulatory effect was produced by polyoxidonium, while the minimum effect was observed for T-activin.
Sigma-Aldrich
Lysozyme from chicken egg white, BioUltra, lyophilized powder, ≥98% (SDS-PAGE), ≥40,000 units/mg protein
Sigma-Aldrich
Lysozyme from chicken egg white, 10 mg/mL
Sigma-Aldrich
Lysozyme from chicken egg white, aseptically filled
Sigma-Aldrich
Lysozyme from chicken egg white, powder or granules, ≥39,000 units/mg protein
Sigma-Aldrich
Lysozyme from chicken egg white, For use as a marker in SDS-PAGE
Sigma-Aldrich
Lysozyme chloride form from chicken egg white, Grade VI, ≥35,000 units/mg protein (E1%/282)
Sigma-Aldrich
Lysozyme from chicken egg white, dialyzed, lyophilized, powder, ~100000 U/mg
Sigma-Aldrich
Lysozyme from chicken egg white, powder (crystalline), 70000-140000 U/mg
Sigma-Aldrich
Lysozyme human, Lysobac, recombinant, expressed in rice, ≥100,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Lysozyme from human neutrophils, ≥95% (SDS-PAGE), lyophilized powder, ≥100,000 units/mg protein (E1%/280)