Merck
  • [The characteristics of chemical composition of content of unicameral bone cysts depending on their growth stage].
[The characteristics of chemical composition of content of unicameral bone cysts depending on their growth stage].
Klinicheskaia laboratornaia diagnostika (2013-01-12)
M V Stogov, S N Luneva, A I Mitrofanov, E A Tkachuk
ABSTRACT
The article deals with the results of study of chemical composition of solitary cysts and blood serum of 27 patients. The results demonstrated that qualitative composition of f content of unicameral bone cysts is identical to chemical composition of blood serum. The results of analysis of total proteolysis activity and acid phosphatase activity in content of cysts can be used as criteria to determine the stage of cyst growth and to evaluate the effectiveness of applied treatment.
Sigma-Aldrich
Phosphatase, Acid from wheat germ, ≥0.4 unit/mg solid
Sigma-Aldrich
Phosphatase, Acid from sweet potato, ammonium sulfate suspension, ≥10.0 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
Phosphatase, Acid from potato, lyophilized powder, ≥0.5 unit/mg solid
Sigma-Aldrich
Phosphatase, Acid from potato, lyophilized powder, ≥3.0 units/mg solid