Merck
  • Ablation of TRIP-Br2, a regulator of fat lipolysis, thermogenesis and oxidative metabolism, prevents diet-induced obesity and insulin resistance.
Ablation of TRIP-Br2, a regulator of fat lipolysis, thermogenesis and oxidative metabolism, prevents diet-induced obesity and insulin resistance.
Nature medicine (2013-01-08)
Chong Wee Liew, Jeremie Boucher, Jit Kong Cheong, Cecile Vernochet, Ho-Jin Koh, Cristina Mallol, Kristy Townsend, Dominique Langin, Dan Kawamori, Jiang Hu, Yu-Hua Tseng, Marc K Hellerstein, Stephen R Farmer, Laurie Goodyear, Alessandro Doria, Matthias Blüher, Stephen I-Hong Hsu, Rohit N Kulkarni
ABSTRACT
Obesity develops as a result of altered energy homeostasis favoring fat storage. Here we describe a new transcription co-regulator for adiposity and energy metabolism, SERTA domain containing 2 (TRIP-Br2, also called SERTAD2). TRIP-Br2-null mice are resistant to obesity and obesity-related insulin resistance. Adipocytes of these knockout mice showed greater stimulated lipolysis secondary to enhanced expression of hormone sensitive lipase (HSL) and β3-adrenergic (Adrb3) receptors. The knockout mice also have higher energy expenditure because of increased adipocyte thermogenesis and oxidative metabolism caused by upregulating key enzymes in their respective processes. Our data show that a cell-cycle transcriptional co-regulator, TRIP-Br2, modulates fat storage through simultaneous regulation of lipolysis, thermogenesis and oxidative metabolism. These data, together with the observation that TRIP-Br2 expression is selectively elevated in visceral fat in obese humans, suggests that this transcriptional co-regulator is a new therapeutic target for counteracting the development of obesity, insulin resistance and hyperlipidemia.
Sigma-Aldrich
Cholesterol Esterase from bovine pancreas, lyophilized powder, ≥200 units/g protein
Sigma-Aldrich
Cholesterol Esterase from Pseudomonas fluorescens, lyophilized powder, ≥10,000 units/g protein
Sigma-Aldrich
Cholesterol Esterase from Pseudomonas sp., lyophilized powder, ≥200,000 units/g protein
Sigma-Aldrich
Cholesterol Esterase from porcine pancreas, lyophilized, powder, white, ~35 U/mg
Sigma-Aldrich
EZ-Magna ChIP® G - Chromatin Immunoprecipitation Kit, Single day chromatin immunoprecipitation (ChIP) kit containing all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions using magnetic G beads. Control primers included.