Merck
  • Identification of a target gene and activating stimulus for the YpdA/YpdB histidine kinase/response regulator system in Escherichia coli.
Identification of a target gene and activating stimulus for the YpdA/YpdB histidine kinase/response regulator system in Escherichia coli.
Journal of bacteriology (2012-12-12)
Luitpold Fried, Stefan Behr, Kirsten Jung
ABSTRACT
Escherichia coli contains 30 two-component systems (TCSs), each consisting of a histidine kinase and a response regulator. Whereas most TCSs are well characterized in this model organism, little is known about the YpdA/YpdB system. To identify YpdB-regulated genes, we compared the transcriptomes of E. coli cells overproducing either YpdB or a control protein. Expression levels of 15 genes differed by more than 1.9-fold between the two strains. A comprehensive evaluation of these genes identified yhjX as the sole target of YpdB. Electrophoretic mobility shift assays with purified YpdB confirmed its interaction with the yhjX promoter. Specifically, YpdB binds to two direct repeats of the motif GGCATTTCAT separated by an 11-bp spacer in the yhjX promoter. yhjX encodes a cytoplasmic membrane protein of unknown function that belongs to the major facilitator superfamily of transporters. Finally, we characterized the pattern of yhjX expression and identified extracellular pyruvate as a stimulus for the YpdA/YpdB system. It is suggested that YpdA/YpdB contributes to nutrient scavenging before entry into stationary phase.
Sigma-Aldrich
Deoxyribonuclease I bovine, recombinant, expressed in Pichia pastoris, buffered aqueous glycerol solution, ≥5,000 units/mg protein
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, lyophilized powder, Protein ≥85 %, ≥400 Kunitz units/mg protein
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Type II-S, lyophilized powder, Protein ≥80 %, ≥2,000 units/mg protein
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Type II, lyophilized powder, Protein ≥80 %, ≥2,000 units/mg protein
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Type IV, lyophilized powder, ≥2,000 Kunitz units/mg protein
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Standardized vial containing 2,000 Kunitz units of DNase I (D4527), vial of ≥0.25 mg total protein
Sigma-Aldrich
Deoxyribonuclease I bovine, recombinant, expressed in Pichia pastoris, lyophilized powder, RNAse and protease, free
Sigma-Aldrich
Deoxyribonuclease I RNase-free solution from bovine pancreas
Sigma-Aldrich
GenElute Gel Extraction Kit, sufficient for 70 purifications