Merck
  • Structural features and antioxidant activity of xylooligosaccharides enzymatically produced from sugarcane bagasse.
Structural features and antioxidant activity of xylooligosaccharides enzymatically produced from sugarcane bagasse.
Bioresource technology (2012-11-08)
Jing Bian, Feng Peng, Xiao-Peng Peng, Pai Peng, Feng Xu, Run-Cang Sun
ABSTRACT
Xylooligosaccharides (XOS) were prepared from xylan-rich hemicelluloses isolated by potassium hydroxide from sugarcane bagasse by hydrolysis with crude xylanase secreted by Pichia stipitis. Hydrolysis for 12h produced XOS with a maximum yield of 31.8%, equivalent to 5.29 mg mL(-1) in the hydrolyzate. XOS with degrees of polymerization (DP) from 2 to 4 (xylobiose, xylotriose, and xylotetraose) were the major components in the hydrolysates, whereas the oligosaccharides with higher DP of 5-6 (xylopentaose and xylohexose) showed a constant low level. FT-IR and NMR ((1)H, (13)C, HSQC) demonstrated that XOS contained Araf and 4-O-Me-α-D-GlcpA residues. The 2,2-diphenyl-1-picrylhydrazyl (DPPH) assay showed that the XOS exhibited concentration-dependent antioxidant activity. The results obtained indicate that the XOS produced from sugarcane bagasse can be employed in food-related applications.
Sigma-Aldrich
Xylanase from Trichoderma viride, lyophilized powder, 100-300 units/mg protein
Supelco
Potassium hydroxide concentrate, 0.1 M KOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Potassium hydroxide, tested according to Ph. Eur.
Supelco
Potassium hydroxide solution, volumetric, 8.0 M KOH (8.0N)
Sigma-Aldrich
Potassium hydroxide, ≥85% KOH basis, pellets, white
Sigma-Aldrich
Potassium hydroxide solution, 45 wt. % in H2O
Sigma-Aldrich
Potassium hydroxide, puriss. p.a., ≥86% (T), pellets
Sigma-Aldrich
Potassium hydroxide, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Sigma-Aldrich
Potassium hydroxide, reagent grade, 90%, flakes
Sigma-Aldrich
Potassium hydroxide, pellets, reag. Ph. Eur., ≥85%
Sigma-Aldrich
Potassium hydroxide, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis
Sigma-Aldrich
Potassium hydroxide, free-flowing, Redi-Dri, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Potassium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, 85-100.5%, pellets
Sigma-Aldrich
Potassium hydroxide, technical, ≥85%, powder