Merck
  • Grape seed proanthocyanidins inhibit colon cancer-induced angiogenesis through suppressing the expression of VEGF and Ang1.
Grape seed proanthocyanidins inhibit colon cancer-induced angiogenesis through suppressing the expression of VEGF and Ang1.
International journal of molecular medicine (2012-10-03)
Shuangsheng Huang, Ninggang Yang, Yuanyuan Liu, Jing Gao, Tao Huang, Lamei Hu, Jin Zhao, Yongquan Li, Caili Li, Xiaosu Zhang
ABSTRACT
Tumor cells trigger angiogenesis through overexpression of various angiogenic factors including vascular endothelial growth factor (VEGF) and angiopoietin 1 (Ang1). Therefore, inhibition of the expression of both VEGF and Ang1, the initial step of tumor angiogenesis, is a promising strategy for cancer chemoprevention and therapy. Grape seed proanthocyanidins (GSPs) are widely consumed dietary supplements that have antitumor activity. Due to their polymeric structure, GSPs are poorly absorbed along the gastrointestinal tract and can reach the colon at high concentrations, allowing these chemicals to act as chemopreventive agents for colon cancer. In the present study, we found that GSPs inhibited colon tumor-induced angiogenesis and, thus, the growth of colon tumor xenografts on the chick chorioallantoic membranes. The mechanisms of their action were related to inhibiting the expression of both VEGF and Ang1 through scavenging reactive oxygen species. Previous studies have demonstrated that the chemopreventive effects of GSPs on colon cancer are associated with their growth inhibitory and apoptosis-inducing effects. Our results demonstrate another mechanism by which GSPs inhibit colon tumor growth, which will be helpful for developing GSPs as a pharmacologically safe angiopreventive agent against colorectal cancer.
Sigma-Aldrich
Ribonuclease A from bovine pancreas, 90%, powder, white, ~80 U/mg
Sigma-Aldrich
Ribonuclease A from bovine pancreas, 4×cryst., ~70 U/mg (acc. to Kunitz)
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type III-A, ≥85% RNase A basis (SDS-PAGE), 85-140 Kunitz units/mg protein
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type I-A, powder, ≥60% RNase A basis (SDS-PAGE), ≥50 Kunitz units/mg protein
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type II-A, ≥60% (SDS-PAGE), >= 60 Kunitz units/mg protein
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type I-AS, 50-100 Kunitz units/mg protein
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type XII-A, ≥90% (SDS-PAGE), 75-125 Kunitz units/mg protein
Sigma-Aldrich
Ribonuclease A from bovine pancreas, for molecular biology, ≥70 Kunitz units/mg protein, lyophilized
Sigma-Aldrich
Ribonuclease B from bovine pancreas, BioReagent, ≥50 Kunitz units/mg protein, ≥80% (SDS-PAGE)
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type X-A, ≥90% (SDS-PAGE), ≥70 Kunitz units/mg protein
Sigma-Aldrich
Ribonuclease A from bovine pancreas, (Solution of 50% glycerol, 10mM Tris-HCL pH 8.0)