Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • The intriguing enhancement of chloroperoxidase mediated one-electron oxidations by azide, a known active-site ligand.
The intriguing enhancement of chloroperoxidase mediated one-electron oxidations by azide, a known active-site ligand.
Biochemical and biophysical research communications (2011-11-15)
Daniel Andrew, Lowell Hager, Kelath Murali Manoj
ABSTRACT
Azide is a well-known inhibitor of heme-enzymes. Herein, we report the counter-intuitive observation that at some concentration regimes, incorporation of azide in the reaction medium enhances chloroperoxidase (CPO, a heme-enzyme) mediated one-electron abstractions from several substrates. A diffusible azidyl radical based mechanism is proposed for explaining the phenomenon. Further, it is projected that the finding could have significant impact on routine in situ or in vitro biochemistry studies involving heme-enzyme systems and azide.
Sigma-Aldrich
Chloroperoxidase from Caldariomyces fumago, aqueous suspension, brown, >10,000 U/mL
Sigma-Aldrich
Chloroperoxidase from Caldariomyces fumago, buffered aqueous suspension, ≥3,000 units/mL
Sigma-Aldrich
Chloroperoxidase from Caldariomyces fumago, buffered aqueous suspension, 1,000-2,000 units/mg protein (E1%/280)