Merck
  • A BEN-domain-containing protein associates with heterochromatin and represses transcription.
A BEN-domain-containing protein associates with heterochromatin and represses transcription.
Journal of cell science (2011-09-15)
Kizhakke M Sathyan, Zhen Shen, Vidisha Tripathi, Kannanganattu V Prasanth, Supriya G Prasanth
ABSTRACT
In eukaryotes, higher order chromatin structure governs crucial cellular processes including DNA replication, transcription and post-transcriptional gene regulation. Specific chromatin-interacting proteins play vital roles in the maintenance of chromatin structure. We have identified BEND3, a quadruple BEN domain-containing protein that is highly conserved amongst vertebrates. BEND3 colocalizes with HP1 and H3 trimethylated at K9 at heterochromatic regions in mammalian cells. Using an in vivo gene locus, we have been able to demonstrate that BEND3 associates with the locus only when it is heterochromatic and dissociates upon activation of transcription. Furthermore, tethering BEND3 inhibits transcription from the locus, indicating that BEND3 is involved in transcriptional repression through its interaction with histone deacetylases and Sall4, a transcription repressor. We further demonstrate that BEND3 is SUMOylated and that such modifications are essential for its role in transcriptional repression. Finally, overexpression of BEND3 causes premature chromatin condensation and extensive heterochromatinization, resulting in cell cycle arrest. Taken together, our data demonstrate the role of a novel heterochromatin-associated protein in transcriptional repression.
Product Description
Millipore
Benzonase® Nuclease, ≥250 units/μL, ≥90% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-PCNA Antibody, clone PC10, clone PC10, Chemicon®, from mouse
Millipore
Human C-Peptide ELISA, This Human C-Peptide ELISA is used to measure & quantify C-Peptide levels in Metabolism & Endocrine research.
Sigma-Aldrich
Anti-HP1α Antibody, clone15.19s2, clone 15.19s2, Upstate®, from mouse
Sigma-Aldrich
Benzonase® Nuclease, ultrapure, ≥250 units/μL, ≥99% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution, ultrapure grade
Sigma-Aldrich
Anti-Heterochromatin Protein-1 β Antibody, clone 1MOD-1A9, ascites fluid, clone 1MOD-1A9, Chemicon®