Merck
  • Ninein is expressed in the cytoplasm of angiogenic tip-cells and regulates tubular morphogenesis of endothelial cells.
Ninein is expressed in the cytoplasm of angiogenic tip-cells and regulates tubular morphogenesis of endothelial cells.
Arteriosclerosis, thrombosis, and vascular biology (2008-09-06)
Taro Matsumoto, Petter Schiller, Lothar C Dieterich, Fuad Bahram, Yuji Iribe, Ulf Hellman, Charlotte Wikner, Gordon Chan, Lena Claesson-Welsh, Anna Dimberg
ABSTRACT
Angiogenesis is an integral part of many physiological processes but may also aggravate pathological conditions such as cancer. Development of effective angiogenesis inhibitors requires a thorough understanding of the molecular mechanisms regulating vessel formation. The aim of this project was to identify proteins that regulate tubular morphogenesis of endothelial cells. Phosphotyrosine-dependent affinity-purification and mass spectrometry showed tyrosine phosphorylation of ninein during tubular morphogenesis of endothelial cells. Ninein was recently identified as a centrosomal microtubule-anchoring protein. Our results show that ninein is localized in the cytoplasm in endothelial cells, and that it is highly expressed in the vasculature in normal and pathological human tissues. Using embryoid bodies as a model of vascular development, we found that ninein is abundantly expressed in the cytoplasm of endothelial cells during sprouting angiogenesis, in particular in the sprouting tip-cell. In accordance, siRNA-dependent silencing of ninein in endothelial cells inhibited tubular morphogenesis. In this study, we show that ninein is expressed in developing vessels and in endothelial tip cells, and that ninein is critical for formation of the vascular tube. These data strongly implicate ninein as an important new regulator of angiogenesis.
Sigma-Aldrich
Duolink® In Situ Mounting Medium with DAPI
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat MINUS, Affinity purified Donkey anti-Goat IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Detection Reagents Red
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Microplate Nuclear Stain, Anti-Fade
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Probemaker PLUS
Sigma-Aldrich
Duolink® In Situ Microplate Heat Transfer Block
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, clone 4G10®, Upstate®, from mouse
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse PLUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents Orange
Sigma-Aldrich
Duolink® In Situ Detection Reagents Green
Sigma-Aldrich
Duolink® In Situ Detection Reagents FarRed
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit MINUS
Sigma-Aldrich
Duolink® In Situ Wash Buffers, Fluorescence
Sigma-Aldrich
Duolink® In Situ Probemaker MINUS
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat PLUS