Merck
  • HPLC method validation for Digitalis and its analogue by pulsed amperometric detection.
HPLC method validation for Digitalis and its analogue by pulsed amperometric detection.
Journal of pharmaceutical and biomedical analysis (2010-08-31)
Ha-Jeong Kwon, Hee-Jung Sim, Sa-im Lee, Yong-Moon Lee, Yong-Duk Park, Seon-Pyo Hong
ABSTRACT
We developed a highly sensitive and selective reversed-phase HPLC-pulsed amperometric detection (RP-HPLC-PAD) method for cardiac glycoside detection. Eight cardiac glycosides were completely separated within 45 min on a reversed-phase column using a water-acetonitrile gradient, and were detected using a PAD under NaOH alkaline conditions. The detection (S/N=3) and quantification (S/N=10) limits for the cardiac glycosides were 0.1-0.3 and 0.3-0.8 ng, respectively. The linear regression coefficient was 0.9962-0.9998 for concentrations of 1-25 μg/mL. Cardiac glycosides in the Digitalis purpurea leaf displayed intra- and inter-day precisions (RSDs) of <9.30% and average recoveries of 98.63-99.94%. The contents of gitoxin, digitonin, and digitoxin in the D. purpurea were 0.197, 0.11, and 0.379 mg/g for leaf dried at 60 °C, 0.058, 0.11, and 0.090 mg/g for leaf dried at ambient temperature, and N.D. (not detected), and 18.379 mg/g, N.D. for seed, respectively. We conclude that our method shows good precision and accuracy.
Sigma-Aldrich
Digitoxin, ≥92% (HPLC), powder
Sigma-Aldrich
Gitoxin, ≥99% (HPLC)
Digitoxin, European Pharmacopoeia (EP) Reference Standard