Merck
  • Decontamination studies with the agents of bovine spongiform encephalopathy and scrapie.
Decontamination studies with the agents of bovine spongiform encephalopathy and scrapie.
Archives of virology (1994-01-01)
D M Taylor, H Fraser, I McConnell, D A Brown, K L Brown, K A Lamza, G R Smith
ABSTRACT
Macerates of bovine brain infected with bovine spongiform encephalopathy (BSE) agent, and rodent brain infected with the 263K or ME7 strains of scrapie agent, were subjected to porous-load autoclaving at temperatures between 134 and 138 degrees C for < or = 60 min. Bioassay in rodents showed that none of the regimens produced complete inactivation. Homogenates of BSE-infected bovine brain were exposed for < or = 120 min to solutions of sodium hypochlorite or sodium dichloroisocyanurate containing < or = 16,500 ppm available chlorine. There was no detectable survival of infectivity after the hypochlorite treatments but none of the dichloroisocyanurate solutions produced complete inactivation. Homogenates of BSE-infected bovine brain, and rodent brain infected with the 263K and ME7 strains of scrapie agent, were exposed for < or = 120 min to 1M or 2M sodium hydroxide but no procedure produced complete inactivation of all agents tested.
Sigma-Aldrich
Alkaline Hypochlorite solution
Pricing and availability is not currently available.