Merck
  • Activated cAMP response element binding protein is overexpressed in human mesotheliomas and inhibits apoptosis.
Activated cAMP response element binding protein is overexpressed in human mesotheliomas and inhibits apoptosis.
The American journal of pathology (2009-10-10)
Arti Shukla, Marcus W Bosenberg, Maximilian B MacPherson, Kelly J Butnor, Nicholas H Heintz, Harvey I Pass, Michele Carbone, Joseph R Testa, Brooke T Mossman
ABSTRACT
Little is known about the cellular mechanisms contributing to the development and chemoresistance of malignant mesothelioma (MM), an aggressive asbestos-associated tumor. A human mesothelial cell line (LP9/TERT-1) and isolated human pleural mesothelial cells showed rapid and protracted asbestos-induced cAMP response element binding protein (CREB1) phosphorylation, which was inhibited in LP9/TERT-1 cells by small molecule inhibitors of epidermal growth factor receptor phosphorylation and protein kinase A. Asbestos increased expression of several CREB target genes (c-FOS, EGR-1, MKP1, BCL2, and MMP13) and apoptosis, which was enhanced using small interfering CREB. Human MM tissue arrays showed elevated endogenous levels of phosphorylated nuclear CREB1 as compared with reactive mesothelial hyperplasias and normal lung tissue. Significantly increased phosphorylated CREB1 and mRNA levels of BCL2, c-FOS, MMP9, and MMP13 were also observed in MM cells in vitro, which were further augmented after addition of Doxorubicin (Dox). Small interfering CREB inhibited migration of MMs, increased apoptosis by Dox, and decreased BCL2 and BCL-xL expression, suggesting a role for these molecules in CREB-induced MM survival. These data indicate that CREB1 and its target genes are up-regulated in asbestos-exposed human mesothelial cells through an epidermal growth factor receptor/protein kinase A pathway. Since activated CREB1 also is increased endogenously in human MM and modifies migration and resistance to Dox-induced apoptosis, inhibition of CREB1 may be a new strategy for MM therapy.
Product Description
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Sodium selenite, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Sodium selenite, γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium selenite, anhydrous, ≥90.0% (RT)
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Sodium selenite, 99%
Sigma-Aldrich
U0126, U0126, CAS 109511-58-2, is a potent and specific inhibitor of MEK1 (IC₅₀ = 72 nM) and MEK2 (IC₅₀ = 58 nM). The inhibition is noncompetitive with respect to both ATP and ERK.
Sigma-Aldrich
KN-93, A cell-permeable, reversible and competitive inhibitor of rat brain CaM kinase II (Ki = 370 nM).
Sigma-Aldrich
AG 1478, A cell-permeable, reversible, ATP-competitive, highly potent and selective inhibitor of epidermal growth factor receptor kinase versus HER2-neu and platelet-derived growth factor receptor kinase.
Sigma-Aldrich
KN-92, KN-92, CAS 1135280-28-2, is useful as a negative control for KN-93 (Cat. No. 422708), a Ca2+/CaM Kinase II inhibitor.