Merck
  • Fibulin-5 suppresses lung cancer invasion by inhibiting matrix metalloproteinase-7 expression.
Fibulin-5 suppresses lung cancer invasion by inhibiting matrix metalloproteinase-7 expression.
Cancer research (2009-07-09)
Wen Yue, Quanhong Sun, Rodney Landreneau, Chuanyue Wu, Jill M Siegfried, Jian Yu, Lin Zhang
ABSTRACT
The high mortality rate of lung cancer is largely due to the spread of disease to other organs. However, the molecular changes driving lung cancer invasion and metastasis remain unclear. In this study, we identified fibulin-5, a vascular ligand for integrin receptors, as a suppressor of lung cancer invasion and metastasis. Fibulin-5 was silenced by promoter hypermethylation in a majority of lung cancer cell lines and primary tumors. It inhibited lung cancer cell invasion and down-regulated matrix metalloproteinase-7 (MMP-7), which promoted lung cancer cell invasion. Knockdown of fibulin-5 was sufficient to stimulate cell invasion and MMP-7 expression. The expression levels of fibulin-5 and MMP-7 were inversely correlated in lung tumors. Suppression of MMP-7 expression by fibulin-5 was mediated by an integrin-binding RGD motif via the extracellular signal-regulated kinase (ERK) pathway. Furthermore, overexpression of fibulin-5 in H460 lung cancer cells inhibited metastasis in mice. Collectively, these results suggest that epigenetic silencing of fibulin-5 promotes lung cancer invasion and metastasis by activating MMP-7 expression through the ERK pathway.
Product Description
Supelco
Crystal Violet, VETRANAL®, analytical standard
Sigma-Aldrich
Crystal Violet, suitable for microscopy
Sigma-Aldrich
Crystal Violet, certified by the BSC
Sigma-Aldrich
Crystal Violet, ACS reagent, ≥90.0% anhydrous basis
Sigma-Aldrich
Crystal Violet, S. No.: 785