Merck
  • Immunization with oligomannose-coated liposome-entrapped dense granule protein 7 protects dams and offspring from Neospora caninum infection in mice.
Immunization with oligomannose-coated liposome-entrapped dense granule protein 7 protects dams and offspring from Neospora caninum infection in mice.
Clinical and vaccine immunology : CVI (2009-04-10)
Yoshifumi Nishikawa, Houshuang Zhang, Yuzuru Ikehara, Naoya Kojima, Xuenan Xuan, Naoaki Yokoyama
ABSTRACT
The present study demonstrates that the subcutaneous administration of Neospora caninum dense granule protein 7 (NcGRA7) entrapped in liposomes coated with mannotriose strongly induces the parasite-specific T-helper type 1 immune response and humoral antibody in mice. Although anti-NcGRA7 immunoglobulin G1 antibody production was induced in mice injected with NcGRA7 alone, the dams and offspring were never protected from N. caninum infection. The immunization of mice with liposome-entrapped NcGRA7 before pregnancy resulted in increased offspring survival and decreased the infection rates in the brains of dams after parasite infection at 6 to 9 days of gestation. In conclusion, oligomannose-coated liposome-entrapped NcGRA7 can be used as a new type of effective vaccine to control neosporosis.
Product Description
SAFC
Minimum Essential Medium, with Earle′s Balanced Salts, with 2.0 mM L-glutamine, without sodium bicarbonate, dry powder, suitable for cell culture
SAFC
Minimum Essential Medium, with Earle′s salts, with 2.0 mM L-glutamine, with 20.0 mM HEPES, with non-essential amino acids, liquid, sterile-filtered, suitable for cell culture
SAFC
Minimum Essential Medium, with Earle′s Balanced Salts, with non-essential amino acids, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
SAFC
Minimum Essential Medium, with Earle′s Balanced Salts, with 2.0 mM L-glutamine, liquid, sterile-filtered, suitable for cell culture
SAFC
Minimum Essential Medium, with Earle′s Balanced Salts, with 25mM HEPES, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
SAFC
Minimum Essential Medium, with Earle′s Balanced Salts, with 2.0 mM L-glutamine, with non-essential amino acids, without sodium bicarbonate, dry powder, suitable for cell culture
SAFC
Minimum Essential Medium, with Earle′s Balanced Salts, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine and sodium bicarbonate. Without arginine, leucine, lysine, and phenol red, liquid, sterile-filtered, suitable for cell culture, designed for isotope labeling for cell culture applications
Sigma-Aldrich
StableCell RPMI-1640, With stable glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
SAFC
RPMI-1640 Medium, with 2.05 mM L-glutamine, with 25mM HEPES, liquid, sterile-filtered, suitable for cell culture
SAFC
RPMI-1640 Medium, HEPES Modification, With 25 mM HEPES, without L-glutamine., liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Modified, with L-glutamine, without phenol red and sodium bicarbonate, powder, suitable for cell culture
SAFC
RPMI-1640 Medium, HEPES Modification, with L-glutamine and 25mM HEPES, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Modified, with sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
SAFC
RPMI-1640 Medium, 10 ×, Without L-glutamine, folic acid and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Dutch Modification, with sodium bicarbonate and 20mM HEPES, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Modified, with sodium bicarbonate, without methionine, cystine and L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Modified, with 20 mM HEPES and L-glutamine, without sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
SAFC
RPMI-1640 Medium, Hybri-Max, Modified, with L-glutamine, 4500 mg/L glucose and 15mM HEPES, without sodium bicarbonate, powder, suitable for hybridoma
SAFC
RPMI-1640 Medium, AutoMod, without L-glutamine and sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine, without glucose and sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture