Merck
  • Dysregulation of FLVCR1a-dependent mitochondrial calcium handling in neural progenitors causes congenital hydrocephalus.
Dysregulation of FLVCR1a-dependent mitochondrial calcium handling in neural progenitors causes congenital hydrocephalus.
Cell reports. Medicine (2024-07-18)
Francesca Bertino, Dibyanti Mukherjee, Massimo Bonora, Christoph Bagowski, Jeannette Nardelli, Livia Metani, Diletta Isabella Zanin Venturini, Diego Chianese, Nicolas Santander, Iris Chiara Salaroglio, Andreas Hentschel, Elisa Quarta, Tullio Genova, Arpana Arjun McKinney, Anna Lucia Allocco, Veronica Fiorito, Sara Petrillo, Giorgia Ammirata, Francesco De Giorgio, Evan Dennis, Garrett Allington, Felicitas Maier, Moneef Shoukier, Karl-Philipp Gloning, Luca Munaron, Federico Mussano, Ettore Salsano, Davide Pareyson, Maja di Rocco, Fiorella Altruda, Georgia Panagiotakos, Kristopher T Kahle, Pierre Gressens, Chiara Riganti, Paolo P Pinton, Andreas Roos, Thomas Arnold, Emanuela Tolosano, Deborah Chiabrando
ABSTRACT
Congenital hydrocephalus (CH), occurring in approximately 1/1,000 live births, represents an important clinical challenge due to the limited knowledge of underlying molecular mechanisms. The discovery of novel CH genes is thus essential to shed light on the intricate processes responsible for ventricular dilatation in CH. Here, we identify FLVCR1 (feline leukemia virus subgroup C receptor 1) as a gene responsible for a severe form of CH in humans and mice. Mechanistically, our data reveal that the full-length isoform encoded by the FLVCR1 gene, FLVCR1a, interacts with the IP3R3-VDAC complex located on mitochondria-associated membranes (MAMs) that controls mitochondrial calcium handling. Loss of Flvcr1a in mouse neural progenitor cells (NPCs) affects mitochondrial calcium levels and energy metabolism, leading to defective cortical neurogenesis and brain ventricle enlargement. These data point to defective NPCs calcium handling and metabolic activity as one of the pathogenetic mechanisms driving CH.
Sigma-Aldrich
Anti-Vinculin antibody, Mouse monoclonal, clone V284, purified from hybridoma cell culture
Sigma-Aldrich
Carbamoylcholine chloride, ≥98% (titration), crystalline
Millipore
Protease Inhibitor Cocktail Set III, Animal-Free, Protease Inhibitor Cocktail Set III, Animal-Free, is a cocktail of six protease inhibitors with broad specificity for the inhibition of aspartic, cysteine & serine proteases & aminopeptidases.
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution
Roche
cOmplete Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Sigma-Aldrich
Anti-SIGMAR1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Histamine dihydrochloride, ≥99% (TLC), powder
Sigma-Aldrich
Phenylmethylsulfonyl Fluoride, Phenylmethylsulfonyl Fluoride, CAS 329-98-6, is an irreversible inhibitor of serine proteases. It causes sulfonylation of the active-site serine residues.
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Tablets provided in EASYpacks