- Protocol to optimize the biobanking of ovarian cancer organoids by accommodating patient-specific differences in stemness potential.
Protocol to optimize the biobanking of ovarian cancer organoids by accommodating patient-specific differences in stemness potential.
STAR protocols (2023-08-16)
Fabian Trillsch, Bastian Czogalla, Fabian Kraus, Alexander Burges, Sven Mahner, Mirjana Kessler
PMID37585293
ABSTRACT
We present a protocol for effective biobanking of epithelial ovarian cancer organoids, considering the heterogeneous clinical presentation and high recurrence rates. Our protocol involves parallel testing of three media to identify patient-specific optimal conditions. We describe steps for tissue dissociation, differential seeding, organoid cultivation, and biobanking. We outline procedures for fixation, embedding, and staining for confocal imaging. Furthermore, we demonstrate that brief cultivation of isolates in 2D on plastic enhances organoid-forming potential in selected lines, expanding their application scope. For complete details on the use and execution of this protocol, please refer to Hoffmann et al.1.
Sigma-Aldrich
TGF-β RI Kinase Inhibitor IV, TGF-β RI Kinase Inhibitor IV, CAS 909910-43-6, is a cell-permeable, selective inhibitor of ALK-4/5/7-mediated signaling (IC₅₀ = 45, 12, and 7.5 nM, respectively).