Merck
  • Effect of Granzyme K, FasL and Interferon-γ Expression in Placentas with Preeclampsia.
Effect of Granzyme K, FasL and Interferon-γ Expression in Placentas with Preeclampsia.
Biomedicines (2024-04-27)
Martina Vukoja, Marina Ćurlin, Katarina Vukojević, Nevenka Jelić-Knezović, Anita Kolobarić, Martina Orlović Vlaho, Violeta Šoljić
ABSTRACT
This study aimed to investigate the cytotoxic activity of decidual lymphocytes and the mRNA/protein expression of cytotoxic proteins in various cell types in the context of preeclampsia (PE) compared to those of healthy pregnancies. We analyzed fresh decidua basalis tissue and tissue embedded in paraffin (FFPE) from PE pregnancies (n = 15) and compared them with those of healthy pregnancies (n = 15) of the corresponding gestational age. Using double immunofluorescence staining, we observed differences in the intensity and distribution of staining for granzyme K (GZMK) and FasL in extravillous trophoblasts. RT-qPCR analysis of FFPE placental tissue showed that GZMK mRNA expression was statistically higher (p < 0.0001) in PE compared to that of healthy controls. On the contrary, there was a low expression (p < 0.001) of FasL mRNA in PE compared to controls, while there was no statistically significant difference for IFN-γ mRNA between PE and controls. Although the level of cytotoxic activity changed depending on the ratio of effector and target cells, there was no significant difference observed between PE and controls in this in vitro study. In conclusion, in PE, extravillous trophoblasts exhibited increased expression of GZMK and decreased expression of FasL. These changes may contribute to impaired trophoblast invasion. However, these alterations did not appear to affect the cytotoxic properties of decidual lymphocytes. Additionally, the possibility of cell sorter separation of decidual lymphocytes would greatly contribute to a better understanding of single cells' genetic profiles.
Sigma-Aldrich
Anti-FAS antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-GZMK antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution