Merck
  • The efficacy of chemotherapy is limited by intratumoral senescent cells expressing PD-L2.
The efficacy of chemotherapy is limited by intratumoral senescent cells expressing PD-L2.
Nature cancer (2024-01-25)
Selim Chaib, José Alberto López-Domínguez, Marta Lalinde-Gutiérrez, Neus Prats, Ines Marin, Olga Boix, Andrea García-Garijo, Kathleen Meyer, María Isabel Muñoz, Mònica Aguilera, Lidia Mateo, Camille Stephan-Otto Attolini, Susana Llanos, Sandra Pérez-Ramos, Marta Escorihuela, Fatima Al-Shahrour, Timothy P Cash, Tamara Tchkonia, James L Kirkland, María Abad, Alena Gros, Joaquín Arribas, Manuel Serrano
ABSTRACT
Chemotherapy often generates intratumoral senescent cancer cells that strongly modify the tumor microenvironment, favoring immunosuppression and tumor growth. We discovered, through an unbiased proteomics screen, that the immune checkpoint inhibitor programmed cell death 1 ligand 2 (PD-L2) is highly upregulated upon induction of senescence in different types of cancer cells. PD-L2 is not required for cells to undergo senescence, but it is critical for senescent cells to evade the immune system and persist intratumorally. Indeed, after chemotherapy, PD-L2-deficient senescent cancer cells are rapidly eliminated and tumors do not produce the senescence-associated chemokines CXCL1 and CXCL2. Accordingly, PD-L2-deficient pancreatic tumors fail to recruit myeloid-derived suppressor cells and undergo regression driven by CD8 T cells after chemotherapy. Finally, antibody-mediated blockade of PD-L2 strongly synergizes with chemotherapy causing remission of mammary tumors in mice. The combination of chemotherapy with anti-PD-L2 provides a therapeutic strategy that exploits vulnerabilities arising from therapy-induced senescence.
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, 1.5-2.0 units/mg solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-PD-L2 Antibody, clone 366C.9E5, clone 366C.12.9E5.E10.19.1F8, from mouse