Merck
  • PRKCSH contributes to TNFSF resistance by extending IGF1R half-life and activation in lung cancer.
PRKCSH contributes to TNFSF resistance by extending IGF1R half-life and activation in lung cancer.
Experimental & molecular medicine (2024-01-11)
Gu-Choul Shin, Hyeong Min Lee, Nayeon Kim, Sang-Uk Seo, Kwang Pyo Kim, Kyun-Hwan Kim
ABSTRACT
Tumor necrosis factor superfamily (TNFSF) resistance contributes to the development and progression of tumors and resistance to various cancer therapies. Tumor-intrinsic alterations involved in the adaptation to the TNFSF response remain largely unknown. Here, we demonstrate that protein kinase C substrate 80K-H (PRKCSH) abundance in lung cancers boosts oncogenic IGF1R activation, leading to TNFSF resistance. PRKCSH abundance is correlated with IGF1R upregulation in lung cancer tissues. Specifically, PRKCSH interacts with IGF1R and extends its half-life. The PRKCSH-IGF1R axis in tumor cells impairs caspase-8 activation, increases Mcl-1 expression, and inhibits caspase-9, leading to an imbalance between cell death and survival. PRKCSH deficiency augmented the antitumor effects of natural killer (NK) cells, representative TNFSF effector cells, in a tumor xenograft IL-2Rg-deficient NOD/SCID (NIG) mouse model. Our data suggest that PRKCSH plays a critical role in TNFSF resistance and may be a potential target to improve the efficacy of NK cell-based cancer therapy.
Sigma-Aldrich
Cycloheximide, ≥95% (HPLC)
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-Mouse IgG (H+L), F(ab′)2 fragment, highly cross adsorbed-Biotin antibody produced in donkey, affinity isolated antibody, lyophilized powder
Sigma-Aldrich
(R)-MG132