Merck
  • Gasdermin D deficiency aborts myeloid calcium influx to drive granulopoiesis in lupus nephritis.
Gasdermin D deficiency aborts myeloid calcium influx to drive granulopoiesis in lupus nephritis.
Cell communication and signaling : CCS (2024-06-04)
Jiani Shen, Feng Li, Xu Han, Dongying Fu, Yiping Xu, Changjian Zhu, Zhou Liang, Ziwen Tang, Ruilin Zheng, Xinrong Hu, Ruoni Lin, Qiaoqiao Pei, Jing Nie, Ning Luo, Xiaoyan Li, Wei Chen, Haiping Mao, Yi Zhou, Xueqing Yu
ABSTRACT
Gasdermin D (GSDMD) is emerging as an important player in autoimmune diseases, but its exact role in lupus nephritis (LN) remains controversial. Here, we identified markedly elevated GSDMD in human and mouse LN kidneys, predominantly in CD11b+ myeloid cells. Global or myeloid-conditional deletion of GSDMD was shown to exacerbate systemic autoimmunity and renal injury in lupus mice with both chronic graft-versus-host (cGVH) disease and nephrotoxic serum (NTS) nephritis. Interestingly, RNA sequencing and flow cytometry revealed that myeloid GSDMD deficiency enhanced granulopoiesis at the hematopoietic sites in LN mice, exhibiting remarkable enrichment of neutrophil-related genes, significant increases in total and immature neutrophils as well as granulocyte/macrophage progenitors (GMPs). GSDMD-deficient GMPs and all-trans-retinoic acid (ATRA)-stimulated human promyelocytes NB4 were further demonstrated to possess enhanced clonogenic and differentiation abilities compared with controls. Mechanistically, GSDMD knockdown promoted self-renewal and granulocyte differentiation by restricting calcium influx, contributing to granulopoiesis. Functionally, GSDMD deficiency led to increased pathogenic neutrophil extracellular traps (NETs) in lupus peripheral blood and bone marrow-derived neutrophils. Taken together, our data establish that GSDMD deletion accelerates LN development by promoting granulopoiesis in a calcium influx-regulated manner, unraveling its unrecognized critical role in LN pathogenesis.
Sigma-Aldrich
Anti-GSDMD antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution