Merck
  • Cytoprotective Effect of Growth Factors Derived From Platelets on Corticosteroid-Treated Primary Anterior Cruciate Ligament-Derived Stromal Cells and Chondrocytes.
Cytoprotective Effect of Growth Factors Derived From Platelets on Corticosteroid-Treated Primary Anterior Cruciate Ligament-Derived Stromal Cells and Chondrocytes.
Cureus (2024-08-28)
Vijay Sharma, Ulka Sakhalkar, Pratiksha Nadkarni, Rashmi Mishal, Dinesh Parandhaman, Kirti Vichare, Anjalina Francis, Mudit Khanna, Mohit Kukreja, Anuka Sharma
ABSTRACT
Background The use of corticosteroids, such as methylprednisolone, for pain management is a common clinical practice. However, it is well known that corticosteroids induce toxicity in anterior cruciate ligament (ACL)-derived stromal cells and chondrocytes. Growth factors from platelets have anti-inflammatory effects that can potentially limit the cytotoxic effects of corticosteroids. In this study, we explored the role of growth factors obtained from the OssinextTM kit (commercially available Wockhardt growth factor concentrate (GFC) kit) in recovering methylprednisolone-induced cell damage. Methodology Primary ACL-derived stromal cells and chondrocytes were isolated from human ligament tissue and articular cartilage, respectively, and characterized by immunophenotyping, gene expression analysis, and immunostaining. GFC obtained from OssinextTM kit was used for the experiments. The ACL-derived stromal cells and chondrocytes were treated with methylprednisolone, alone or in combination with GFC. Cell viability was measured by the neutral red uptake assay. Changes in cell morphology and collagen pattern were observed microscopically by H&E staining and immunostaining, respectively. Cell proliferation was assessed by cell migration assay, and the cell ultra-structure was analyzed using transmission electron microscopy. Results Methylprednisolone was found to induce cytotoxicity, altered cell morphology, reduced cell proliferation, and organelle damage in both ACL-derived stromal cells and chondrocytes. GFC obtained from the OssinextTM kit was able to restore cell viability and reverse the cell structure damages induced by methylprednisolone. GFC was found to recover and protect the cells, both when used in combination with steroids and when used after the steroid treatment. Conclusions The results indicate that GFC may be clinically beneficial when used in combination with steroids to mitigate their adverse effects.
Sigma-Aldrich
Anti-TNC antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution