Merck
  • Splicing Factor PQBP1 Curtails BAX Expression to Promote Ovarian Cancer Progression.
Splicing Factor PQBP1 Curtails BAX Expression to Promote Ovarian Cancer Progression.
Advanced science (Weinheim, Baden-Wurttemberg, Germany) (2024-02-12)
Xihan Liu, Jiaojiao Zhang, Zixiang Wang, Mingyao Yan, Meining Xu, Gaoyuan Li, Victoria Shender, Jian-Jun Wei, Jianqiao Li, Changshun Shao, Shiqian Zhang, Beihua Kong, Kun Song, Zhaojian Liu
ABSTRACT
Splicing factor polyglutamine binding protein-1 (PQBP1) is abundantly expressed in the central nervous system during development, and mutations in the gene cause intellectual disability. However, the roles of PQBP1 in cancer progression remain largely unknown. Here, it is shown that PQBP1 overexpression promotes tumor progression and indicates worse prognosis in ovarian cancer. Integrative analysis of spyCLIP-seq and RNA-seq data reveals that PQBP1 preferentially binds to exon regions and modulates exon skipping. Mechanistically, it is shown that PQBP1 regulates the splicing of genes related to the apoptotic signaling pathway, including BAX. PQBP1 promotes BAX exon 2 skipping to generate a truncated isoform that undergoes degradation by nonsense-mediated mRNA decay, thus making cancer cells resistant to apoptosis. In contrast, PQBP1 depletion or splice-switching antisense oligonucleotides promote exon 2 inclusion and thus increase BAX expression, leading to inhibition of tumor growth. Together, the results demonstrate an oncogenic role of PQBP1 in ovarian cancer and suggest that targeting the aberrant splicing mediated by PQBP1 has therapeutic potential in cancer treatment.
Sigma-Aldrich
Anti-PQBP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.